missing translation for 'onlineSavingsMsg'
Learn More

3-n-Hexadecylthiophene, 97%

CAS: 119269-24-8 | C20H36S | 308.57 g/mol

$266.56 - $806.74

Chemical Identifiers

CAS 119269-24-8
Molecular Formula C20H36S
Molecular Weight (g/mol) 308.57
MDL Number MFCD07368973
InChI Key FRVZSODZVJPMKO-UHFFFAOYSA-N
Synonym 3-n-hexadecylthiophene, thiophene, 3-hexadecyl, 3-hexadecyl-thiophene, ksc173e6j, acmc-1c275
PubChem CID 2769638
IUPAC Name 3-hexadecylthiophene
SMILES CCCCCCCCCCCCCCCCC1=CSC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2737103
View Documents
Thermo Scientific Chemicals
H2737103
1 g
Each for $266.56
Only null left
 
AAH2737106
View Documents
Thermo Scientific Chemicals
H2737106
5 g
Each for $806.74
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

119269-24-8
308.57
FRVZSODZVJPMKO-UHFFFAOYSA-N
2769638
CCCCCCCCCCCCCCCCC1=CSC=C1
C20H36S
MFCD07368973
3-n-hexadecylthiophene, thiophene, 3-hexadecyl, 3-hexadecyl-thiophene, ksc173e6j, acmc-1c275
3-hexadecylthiophene

Specifications

119269-24-8
>110°C (230°F)
MFCD07368973
3-n-hexadecylthiophene, thiophene, 3-hexadecyl, 3-hexadecyl-thiophene, ksc173e6j, acmc-1c275
CCCCCCCCCCCCCCCCC1=CSC=C1
308.57
308.57
3-n-Hexadecylthiophene
27°C to 32°C
C20H36S
1 g
FRVZSODZVJPMKO-UHFFFAOYSA-N
3-hexadecylthiophene
2769638
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only