missing translation for 'onlineSavingsMsg'
Learn More

3-(N-Ethyl-3-methylanilino)-2-hydroxypropanesulfonic acid sodium salt, Spectrophotometric Det. of H2O2, MilliporeSigma™ Supelco™

Catalog No. 111015631
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015631 5 g
111015630 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015631 Supplier Merck Emd Millipore Supplier No. E86315G
Only null left

Chemical Identifiers

MDL Number MFCD00040641
Synonym 3-(N-Ethyl-m-toluidino)-2-hydroxypropanesulfonic acid sodium salt; N-Ethyl-N-(2-hydroxy-3-sulfopropyl)-m-toluidine sodium salt; Sodium 3-(N-ethyl-3-methylanilino)-2-hydroxypropanesulfonate; Sodium N-ethyl-N-(2-hydroxy-3-sulfopropyl)-m-toluidine; TOOS

Specifications

CAS 82692-93-1
Packaging Poly Bottle
Linear Formula C12H18 NNaO4 S
Quantity 5 g
Formula Weight 295.33
Physical Form Neat