missing translation for 'onlineSavingsMsg'
Learn More

3-(N-Ethyl-3-methylanilino)-2-hydroxypropanesulfonic acid sodium salt,Spectrophotometric Det. of H2O2, ≥98.0%, MilliporeSigma™ Supelco™

Catalog No. 111015629
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015629 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015629 Supplier Merck Emd Millipore Supplier No. 04340500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00040641
Synonym 3-(N-Ethyl-m-toluidino)-2-hydroxypropanesulfonic acid sodium salt; N-Ethyl-N-(2-hydroxy-3-sulfopropyl)-m-toluidine sodium salt; Sodium 3-(N-ethyl-3-methylanilino)-2-hydroxypropanesulfonate; Sodium N-ethyl-N-(2-hydroxy-3-sulfopropyl)-m-toluidine; TOOS

Specifications

CAS 82692-93-1
Linear Formula C12H18 NNaO4 S
Quantity 500 mg
Formula Weight 295.33
Percent Purity ≥98.0% (dried material, NT); ≥98.0%