missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Morpholinone 98.0+%, TCI America™
Supplier: TCI America M278825G
Specifications
| 3-Morpholinone | |
| 105°C | |
| 142°C | |
| MFCD00631009 | |
| 3-morpholinone, 3-ketomorpholine, morpholinone, morpholine-3-one, 3-oxomorpholine, 3-oxo-morpholine, acmc-1bpa5, ksc180o3n, morpholin-3-one; 3-morpholone | |
| O=C1COCCN1 | |
| 101.11 | |
| 101.11 | |
| Crystalline Powder |
| 109-11-5 | |
| White | |
| C4H7NO2 | |
| 25 g | |
| VSEAAEQOQBMPQF-UHFFFAOYSA-N | |
| morpholin-3-one | |
| 66953 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 109-11-5 | |
| 101.11 | |
| VSEAAEQOQBMPQF-UHFFFAOYSA-N | |
| 66953 | |
| O=C1COCCN1 |
| C4H7NO2 | |
| MFCD00631009 | |
| 3-morpholinone, 3-ketomorpholine, morpholinone, morpholine-3-one, 3-oxomorpholine, 3-oxo-morpholine, acmc-1bpa5, ksc180o3n, morpholin-3-one; 3-morpholone | |
| morpholin-3-one |
Safety and Handling
TSCA : No