missing translation for 'onlineSavingsMsg'
Learn More

3-Morpholinone 98.0+%, TCI America™

Catalog No. p-7133428
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27885G 5 g
M278825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27885G Supplier TCI America Supplier No. M27885G
Only null left

Chemical Identifiers

CAS 109-11-5
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.11
MDL Number MFCD00631009
InChI Key VSEAAEQOQBMPQF-UHFFFAOYSA-N
Synonym 3-morpholinone, 3-ketomorpholine, morpholinone, morpholine-3-one, 3-oxomorpholine, 3-oxo-morpholine, acmc-1bpa5, ksc180o3n, morpholin-3-one; 3-morpholone
PubChem CID 66953
IUPAC Name morpholin-3-one
SMILES O=C1COCCN1

Specifications

Melting Point 105°C
Color White
Boiling Point 142°C
Quantity 5 g
Formula Weight 101.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Morpholinone