missing translation for 'onlineSavingsMsg'
Learn More

3-Methylxanthine 98.0+%, TCI America™

Catalog No. p-7133005
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20735G 5 g
M207325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20735G Supplier TCI America Supplier No. M20735G
Only null left

Chemical Identifiers

CAS 1076-22-8
Molecular Formula C6H6N4O2
Molecular Weight (g/mol) 166.14
MDL Number MFCD00005580
InChI Key GMSNIKWWOQHZGF-UHFFFAOYSA-N
Synonym 3-methylxanthine, 3-methyl-1h-purine-2,6 3h,7h-dione, 2,6-dihydroxy-3-methylpurine, xanthine, 3-methyl, 3 mx, 3-methyl xanthine, 1h-purine-2,6-dione, 3,7-dihydro-3-methyl, unii-ws6x982oec, 3-methyl-2,3,6,7-tetrahydro-1h-purine-2,6-dione, ccris 5817
PubChem CID 70639
ChEBI CHEBI:62207
IUPAC Name 3-methyl-7H-purine-2,6-dione
SMILES CN1C2=C(C(=O)NC1=O)NC=N2

Specifications

Melting Point 360°C
Color White-Yellow
Quantity 5 g
Formula Weight 166.14
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methylxanthine
RTECSNumber ZD8750000
TSCA No