accessibility menu, dialog, popup

UserName

CAS RN 1076-22-8

IUPAC Name: 3-methyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Synonyms: 3-methylxanthine N(1)-demethyltheophylline
Molecular Weight (g/mol): 166.14
Molecular Formula: C6H6N4O2
InChi Key: GMSNIKWWOQHZGF-UHFFFAOYSA-N
SMILES: CN1C2=C(NC=N2)C(=O)NC1=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)