missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methylxanthine, 98+%
Description
3-Methylxanthine is a Xanthine derivative with diuretic, cardiac stimulant and smooth muscle relaxant activities; isomeric with theobromine.They can also act as Bronchodilator.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Methylxanthine is a Xanthine derivative with diuretic, cardiac stimulant and smooth muscle relaxant activities; isomeric with theobromine.They can also act as Bronchodilator.
Solubility
Insoluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
3-Methylxanthine is a Xanthine derivative with diuretic, cardiac stimulant and smooth muscle relaxant activities; isomeric with theobromine.They can also act as Bronchodilator.
Solubility
Insoluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Specifications
| Melting Point | >300°C |
| Quantity | 250 mg |
| Solubility Information | Insoluble in water. |
| Formula Weight | 166.14 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 3-Methylxanthine |
Safety and Handling
| Hazard Category | H302 |
| Precautionary Statement | P264b-P270-P301+P312-P330-P501c |
| EINECSNumber | 214-058-1 |
| RTECSNumber | ZD8750000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only