missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylthio)benzoic Acid 98.0+%, TCI America™

Catalog No. M18531G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M18531G 1 g
M18535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M18531G Supplier TCI America Supplier No. M18531G

Chemical Identifiers

CAS 825-99-0
Molecular Formula C8H8O2S
Molecular Weight (g/mol) 168.21
MDL Number MFCD00156993
InChI Key PZGADOOBMVLBJE-UHFFFAOYSA-N
Synonym 3-methylthio benzoic acid, 3-methylsulfanyl benzoic acid, benzoic acid, 3-methylthio, benzoic acid, m-methylthio, 3-mercaptobenzoic acid s-methyl ether, 3-methylsulfanyl-benzoic acid, 3-carboxythioanisole, pubchem10679
PubChem CID 220329
IUPAC Name 3-methylsulfanylbenzoic acid
SMILES CSC1=CC=CC(=C1)C(=O)O

Specifications

Melting Point 128°C
Color White-Yellow
Quantity 1 g
Formula Weight 168.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Methylthio)benzoic Acid