missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylthio)aniline, 97%

Catalog No. AAB2318406
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2318406 5 g
AAB2318414 25 g
AAB2318422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2318406 Supplier Thermo Scientific Chemicals Supplier No. B2318406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1783-81-9 | C7H9NS | 139.216 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1783-81-9
Molecular Formula C7H9NS
Molecular Weight (g/mol) 139.216
MDL Number MFCD00007793
InChI Key KCHLDNLIJVSRPK-UHFFFAOYSA-N
Synonym 3-methylthio aniline, 3-aminothioanisole, m-aminothioanisole, m-methylthio aniline, benzenamine, 3-methylthio, 3-methylmercapto aniline, 3-methylsulfanyl aniline, 3-methylmercaptoaniline, 3-methylthiobenzenamine, m-aminophenyl methyl sulfide
PubChem CID 15693
IUPAC Name 3-methylsulfanylaniline
SMILES CSC1=CC=CC(=C1)N

Specifications

Density 1.13
Boiling Point 163°C to 165°C (16 mmHg)
Flash Point >110°C (230°F)
Odor Unpleasant
Refractive Index 1.637
Quantity 5 g
UN Number UN2810
Beilstein 2078599
Formula Weight 139.23
Percent Purity 97%
Chemical Name or Material 3-(Methylthio)aniline
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 217-262-5
RTECSNumber BY6301000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only