missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Methylthio)-1-hexanol 98.0+%, TCI America™

$70.77 - $189.28

Chemical Identifiers

CAS 51755-66-9
Molecular Formula C7H16OS
Molecular Weight (g/mol) 148.264
MDL Number MFCD00010674
InChI Key JSASXSHMJYRPCM-UHFFFAOYSA-N
Synonym 1-hexanol, 3-methylthio, 3-methylmercapto-1-hexanol, 3-methylsulfanyl hexan-1-ol, 3-methylthio hexan-1-ol, 3-methylthio hexanol, 3-methylthio-1-hexanol, 3-methylthiohexanol, fema no. 3438, r-3-methylthio-1-hexanol
PubChem CID 65413
IUPAC Name 3-methylsulfanylhexan-1-ol
SMILES CCCC(CCO)SC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M20505G View Documents Tci America
M20505G
5g Each for $70.77
 
M205025G View Documents Tci America
M205025G
25g Each for $189.28
 
Specifications

Chemical Identifiers

51755-66-9
148.264
JSASXSHMJYRPCM-UHFFFAOYSA-N
65413
CCCC(CCO)SC
C7H16OS
MFCD00010674
1-hexanol, 3-methylthio, 3-methylmercapto-1-hexanol, 3-methylsulfanyl hexan-1-ol, 3-methylthio hexan-1-ol, 3-methylthio hexanol, 3-methylthio-1-hexanol, 3-methylthiohexanol, fema no. 3438, r-3-methylthio-1-hexanol
3-methylsulfanylhexan-1-ol

Specifications

62°C
5g
C7H16OS
1-hexanol, 3-methylthio, 3-methylmercapto-1-hexanol, 3-methylsulfanyl hexan-1-ol, 3-methylthio hexan-1-ol, 3-methylthio hexanol, 3-methylthio-1-hexanol, 3-methylthiohexanol, fema no. 3438, r-3-methylthio-1-hexanol
CCCC(CCO)SC
148.264
148.26
≥98.0% (GC)
Yellow
51755-66-9
MFCD00010674
JSASXSHMJYRPCM-UHFFFAOYSA-N
3-methylsulfanylhexan-1-ol
65413
Clear Liquid at 20°C
3-(Methylthio)-1-hexanol
Safety and Handling

Safety and Handling

TSCA : No