Learn More
3-(Methylsulfonyl)benzeneboronic acid, 98%
CAS: 373384-18-0 | C7H9BO4S | 200.015 g/mol
$222.42 - $958.56
Chemical Identifiers
| CAS | 373384-18-0 |
|---|---|
| Molecular Formula | C7H9BO4S |
| Molecular Weight (g/mol) | 200.015 |
| MDL Number | MFCD03092935 |
| InChI Key | HZFFUMBZBGETES-UHFFFAOYSA-N |
| Synonym | 3-methylsulfonyl phenylboronic acid, 3-methylsulfonyl phenyl boronic acid, 3-methylsulfonylphenyl boronic acid, 3-methanesulfonyl phenylboronic acid, 3-methanesulfonyl phenyl boronic acid, 3-methanesulfonylphenylboronic acid, boronic acid, 3-methylsulfonyl phenyl, 3-methanesulfonylphenyl boronic acid, 3-methanesulphonyl phenylboronic acid |
| PubChem CID | 2773542 |
| IUPAC Name | (3-methylsulfonylphenyl)boronic acid |
| SMILES | B(C1=CC(=CC=C1)S(=O)(=O)C)(O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5291603
|
Thermo Scientific Chemicals
H5291603 |
1 g |
Each for $222.42
|
|
|||||
|
AAH5291606
|
Thermo Scientific Chemicals
H5291606 |
5 g |
Each for $958.56
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 373384-18-0 | |
| 200.015 | |
| HZFFUMBZBGETES-UHFFFAOYSA-N | |
| 2773542 | |
| B(C1=CC(=CC=C1)S(=O)(=O)C)(O)O |
| C7H9BO4S | |
| MFCD03092935 | |
| 3-methylsulfonyl phenylboronic acid, 3-methylsulfonyl phenyl boronic acid, 3-methylsulfonylphenyl boronic acid, 3-methanesulfonyl phenylboronic acid, 3-methanesulfonyl phenyl boronic acid, 3-methanesulfonylphenylboronic acid, boronic acid, 3-methylsulfonyl phenyl, 3-methanesulfonylphenyl boronic acid, 3-methanesulphonyl phenylboronic acid | |
| (3-methylsulfonylphenyl)boronic acid |
Specifications
| 373384-18-0 | |
| C7H9BO4S | |
| 1 g | |
| HZFFUMBZBGETES-UHFFFAOYSA-N | |
| (3-methylsulfonylphenyl)boronic acid | |
| 2773542 | |
| 98% |
| 102°C to 110°C | |
| MFCD03092935 | |
| 3-methylsulfonyl phenylboronic acid, 3-methylsulfonyl phenyl boronic acid, 3-methylsulfonylphenyl boronic acid, 3-methanesulfonyl phenylboronic acid, 3-methanesulfonyl phenyl boronic acid, 3-methanesulfonylphenylboronic acid, boronic acid, 3-methylsulfonyl phenyl, 3-methanesulfonylphenyl boronic acid, 3-methanesulphonyl phenylboronic acid | |
| B(C1=CC(=CC=C1)S(=O)(=O)C)(O)O | |
| 200.015 | |
| 200.02 | |
| 3-(Methylsulfonyl)benzeneboronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only