missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylsulfonyl)benzeneboronic acid, 98%

Catalog No. AAH5291606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5291606 5 g
AAH5291603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5291606 Supplier Thermo Scientific Chemicals Supplier No. H5291606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 373384-18-0 | C7H9BO4S | 200.015 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 373384-18-0
Molecular Formula C7H9BO4S
Molecular Weight (g/mol) 200.015
MDL Number MFCD03092935
InChI Key HZFFUMBZBGETES-UHFFFAOYSA-N
Synonym 3-methylsulfonyl phenylboronic acid, 3-methylsulfonyl phenyl boronic acid, 3-methylsulfonylphenyl boronic acid, 3-methanesulfonyl phenylboronic acid, 3-methanesulfonyl phenyl boronic acid, 3-methanesulfonylphenylboronic acid, boronic acid, 3-methylsulfonyl phenyl, 3-methanesulfonylphenyl boronic acid, 3-methanesulphonyl phenylboronic acid
PubChem CID 2773542
IUPAC Name (3-methylsulfonylphenyl)boronic acid
SMILES B(C1=CC(=CC=C1)S(=O)(=O)C)(O)O

Specifications

Melting Point 102°C to 110°C
Quantity 5 g
Formula Weight 200.02
Percent Purity 98%
Chemical Name or Material 3-(Methylsulfonyl)benzeneboronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only