missing translation for 'onlineSavingsMsg'
Learn More

3-Methylsalicylic Acid 98.0+%, TCI America™

Supplier:  TCI America C0408500G

 View more versions of this product

Catalog No. C0408500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Methylsalicylic Acid
167°C
C8H8O3
500 g
WHSXTWFYRGOBGO-UHFFFAOYSA-N
2-hydroxy-3-methylbenzoic acid
6738
152.15
Crystalline Powder
83-40-9
Red-White
MFCD00002448
3-methylsalicylic acid, hydroxytoluic acid, o-cresotic acid, o-cresotinic acid, cresotic acid, o-homosalicylic acid, 2-hydroxy-m-toluic acid, 2,3-cresotinic acid, cresotinic acid, benzoic acid, 2-hydroxy-3-methyl
CC1=CC=CC(C(O)=O)=C1O
152.15
CHEBI:20141
≥98.0% (GC,T)

Chemical Identifiers

83-40-9
152.15
WHSXTWFYRGOBGO-UHFFFAOYSA-N
6738
2-hydroxy-3-methylbenzoic acid
C8H8O3
MFCD00002448
3-methylsalicylic acid, hydroxytoluic acid, o-cresotic acid, o-cresotinic acid, cresotic acid, o-homosalicylic acid, 2-hydroxy-m-toluic acid, 2,3-cresotinic acid, cresotinic acid, benzoic acid, 2-hydroxy-3-methyl
CHEBI:20141
CC1=CC=CC(C(O)=O)=C1O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1631

RTECSNumber : GP3920000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions