missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methylsalicylic Acid 98.0+%, TCI America™
Supplier: TCI America C0408100G
Specifications
| 3-Methylsalicylic Acid | |
| 167°C | |
| C8H8O3 | |
| 100 g | |
| WHSXTWFYRGOBGO-UHFFFAOYSA-N | |
| 2-hydroxy-3-methylbenzoic acid | |
| 6738 | |
| 152.15 | |
| Crystalline Powder |
| 83-40-9 | |
| Red-White | |
| MFCD00002448 | |
| 3-methylsalicylic acid, hydroxytoluic acid, o-cresotic acid, o-cresotinic acid, cresotic acid, o-homosalicylic acid, 2-hydroxy-m-toluic acid, 2,3-cresotinic acid, cresotinic acid, benzoic acid, 2-hydroxy-3-methyl | |
| CC1=CC=CC(C(O)=O)=C1O | |
| 152.15 | |
| CHEBI:20141 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 83-40-9 | |
| 152.15 | |
| WHSXTWFYRGOBGO-UHFFFAOYSA-N | |
| 6738 | |
| 2-hydroxy-3-methylbenzoic acid |
| C8H8O3 | |
| MFCD00002448 | |
| 3-methylsalicylic acid, hydroxytoluic acid, o-cresotic acid, o-cresotinic acid, cresotic acid, o-homosalicylic acid, 2-hydroxy-m-toluic acid, 2,3-cresotinic acid, cresotinic acid, benzoic acid, 2-hydroxy-3-methyl | |
| CHEBI:20141 | |
| CC1=CC=CC(C(O)=O)=C1O |
Safety and Handling
EINECSNumber : (3)-1631
RTECSNumber : GP3920000
TSCA : Yes