missing translation for 'onlineSavingsMsg'
Learn More

3-Methylsalicylic Acid 98.0+%, TCI America™

Catalog No. C040825G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C040825G 25 g
C0408100G 100 g
C0408500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C040825G Supplier TCI America Supplier No. C040825G

Chemical Identifiers

CAS 83-40-9
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00002448
InChI Key WHSXTWFYRGOBGO-UHFFFAOYSA-N
Synonym 3-methylsalicylic acid, hydroxytoluic acid, o-cresotic acid, o-cresotinic acid, cresotic acid, o-homosalicylic acid, 2-hydroxy-m-toluic acid, 2,3-cresotinic acid, cresotinic acid, benzoic acid, 2-hydroxy-3-methyl
PubChem CID 6738
ChEBI CHEBI:20141
IUPAC Name 2-hydroxy-3-methylbenzoic acid
SMILES CC1=CC=CC(C(O)=O)=C1O

Specifications

Melting Point 167°C
Color Red-White
Quantity 25 g
Formula Weight 152.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methylsalicylic Acid
EINECSNumber (3)-1631
RTECSNumber GP3920000
TSCA Yes