missing translation for 'onlineSavingsMsg'
Learn More

3-Methylsalicylic acid, 99%

Catalog No. AAL1582522
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1582522 100 g
AAL1582514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1582522 Supplier Thermo Scientific Chemicals Supplier No. L1582522
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 83-40-9 | C8H8O3 | 152.15 g/mol

3-Methylsalicylic acid was used in the synthesis of 8-methyl-2-thioxo-2,3-dihydro-4H-1,3-benzoxazin-4-one, 7-hydroxy-8-methyl-2-thioxo-2,3-dihydro-4H-1,3-benzoxazin-4-one, (R)-, (S)- and (RS)-hydroxymethylmexiletine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methylsalicylic acid was used in the synthesis of 8-methyl-2-thioxo-2,3-dihydro-4H-1,3-benzoxazin-4-one, 7-hydroxy-8-methyl-2-thioxo-2,3-dihydro-4H-1,3-benzoxazin-4-one, (R)-, (S)- and (RS)-hydroxymethylmexiletine.

Solubility
Soluble in Alcohol, Chloroform, Ether and Water (hot).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 83-40-9
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00002448
InChI Key WHSXTWFYRGOBGO-UHFFFAOYSA-N
Synonym 3-methylsalicylic acid, hydroxytoluic acid, o-cresotic acid, o-cresotinic acid, cresotic acid, o-homosalicylic acid, 2-hydroxy-m-toluic acid, 2,3-cresotinic acid, cresotinic acid, benzoic acid, 2-hydroxy-3-methyl
PubChem CID 6738
ChEBI CHEBI:20141
IUPAC Name 2-hydroxy-3-methylbenzoic acid
SMILES CC1=CC=CC(C(O)=O)=C1O

Specifications

Melting Point 163°C to 167°C
Odor Odorless
Quantity 100 g
Beilstein 2086811
Merck Index 14,2581
Solubility Information Soluble in Alcohol,Chloroform,Ether and Water (hot).
Formula Weight 152.15
Percent Purity 99%
Chemical Name or Material 3-Methylsalicylic acid
Hazard Category H302-H315-H318-H335
Hazard Statement GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
EINECSNumber 201-473-8
RTECSNumber GP3920000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only