missing translation for 'onlineSavingsMsg'
Learn More

3-Methylquinoline N-oxide, 97%, Thermo Scientific Chemicals

Catalog No. AAL1063503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1063503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1063503 Supplier Thermo Scientific Chemicals Supplier No. L1063503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1873-55-8
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.188
MDL Number MFCD00082650
InChI Key CBGUMTBCWWMBRV-UHFFFAOYSA-N
Synonym 3-methylquinoline n-oxide, 3-methylquinoline-n-oxide, 3-methylquinoline 1-oxide, 3-methylquinolin-1-ium-1-olate, 3-methyl-quinoline-1-oxide, 3-methylquinolin-1-ol, acmc-1cfdi, qnmo01, 3-methyl-1-azanaphthalene n-oxide, 3-methyl-1-oxidanidyl-quinolin-1-ium
PubChem CID 7021466
IUPAC Name 3-methyl-1-oxidoquinolin-1-ium
SMILES CC1=CC2=CC=CC=C2[N+](=C1)[O-]

Specifications

Melting Point 88°C to 89°C
Quantity 1 g
Beilstein 118125
Formula Weight 159.19
Percent Purity 97%
Chemical Name or Material 3-Methylquinoline N-oxide
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only