accessibility menu, dialog, popup

UserName

H3CNO

CAS RN 1873-55-8

IUPAC Name: 3-methylquinolin-1-ium-1-olate
Synonyms: 3-methylquinolin-1-ium-1-olate
Molecular Weight (g/mol): 159.19
Molecular Formula: C10H9NO
InChi Key: CBGUMTBCWWMBRV-UHFFFAOYSA-N
SMILES: CC1=C[N+]([O-])=C2C=CC=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results