missing translation for 'onlineSavingsMsg'
Learn More

3-Methylpyridine-2-thiocarboxamide, 97%, Thermo Scientific Chemicals

Catalog No. AAH5179403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5179403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5179403 Supplier Thermo Scientific Chemicals Supplier No. H5179403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 334017-95-7 | Molecular Formula: C7H8N2S | Molecular Weight: 152.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 334017-95-7
Molecular Formula C7H8N2S
Molecular Weight (g/mol) 152.22
MDL Number MFCD12026307
InChI Key PEENLLJOQZUCDN-UHFFFAOYSA-N
Synonym 3-methylpyridine-2-thiocarboxamide, 3-methylpyridine-2-carbothioic acid amide, 2-pyridinecarbothioamide, 3-methyl, 3-methylpyridine-2-thioamide
PubChem CID 22494211
IUPAC Name 3-methylpyridine-2-carbothioamide
SMILES CC1=C(N=CC=C1)C(N)=S

Specifications

Melting Point 117°C to 122°C
Quantity 1 g
Formula Weight 152.21
Percent Purity 97%
Chemical Name or Material 3-Methylpyridine-2-thiocarboxamide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only