CAS RN 334017-95-7
IUPAC Name:
3-methylpyridine-2-carbothioamide
Synonyms:
3-methylpyridine-2-carbothioamide
Molecular Weight (g/mol):
152.22
Molecular Formula:
C7H8N2S
InChi Key:
PEENLLJOQZUCDN-UHFFFAOYSA-N
SMILES:
CC1=C(N=CC=C1)C(N)=S
1
–
1
of
1
results
1
Promotions Available
3-Methylpyridine-2-thiocarboxamide, 97%, Thermo Scientific Chemicals
3-Methylpyridine-2-thiocarboxamide, 97%, Thermo Scientific Chemicals
CAS: 334017-95-7 Molecular Formula: C7H8N2S Molecular Weight (g/mol): 152.22 MDL Number: MFCD12026307 InChI Key: PEENLLJOQZUCDN-UHFFFAOYSA-N Synonym: 3-methylpyridine-2-thiocarboxamide,3-methylpyridine-2-carbothioic acid amide,2-pyridinecarbothioamide, 3-methyl,3-methylpyridine-2-thioamide PubChem CID: 22494211 IUPAC Name: 3-methylpyridine-2-carbothioamide SMILES: CC1=C(N=CC=C1)C(N)=S