missing translation for 'onlineSavingsMsg'
Learn More

3-Methylpyrazole-4-carboxaldehyde 98.0+%, TCI America™

Supplier:  TCI America M2643200MG

 View more versions of this product

Catalog No. M2643200MG


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Methylpyrazole-4-carboxaldehyde
110°C
C5H6N2O
200 mg
NWDMGTFNIOCVDU-UHFFFAOYSA-N
5-methyl-1H-pyrazole-4-carbaldehyde
2760056
≥98.0% (GC)
112758-40-4
White-Yellow
MFCD01313822
3-methyl-1h-pyrazole-4-carbaldehyde, 3-methyl-1h-pyrazole-4-carboxaldehyde, 3-methylpyrazole-4-carboxaldehyde, 3-methylpyrazole-4-carbaldehyde, 1h-pyrazole-4-carboxaldehyde, 3-methyl, 3-methyl-2h-pyrazole-4-carbaldehyde, 5-methylpyrazole-4-carbaldehyde, pubchem16065, acmc-1brby, 4-formyl-3-methylpyrazole
CC1=C(C=O)C=NN1
110.12
110.12
Crystalline Powder

Chemical Identifiers

112758-40-4
110.12
NWDMGTFNIOCVDU-UHFFFAOYSA-N
2760056
CC1=C(C=O)C=NN1
C5H6N2O
MFCD01313822
3-methyl-1h-pyrazole-4-carbaldehyde, 3-methyl-1h-pyrazole-4-carboxaldehyde, 3-methylpyrazole-4-carboxaldehyde, 3-methylpyrazole-4-carbaldehyde, 1h-pyrazole-4-carboxaldehyde, 3-methyl, 3-methyl-2h-pyrazole-4-carbaldehyde, 5-methylpyrazole-4-carbaldehyde, pubchem16065, acmc-1brby, 4-formyl-3-methylpyrazole
5-methyl-1H-pyrazole-4-carbaldehyde
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions