missing translation for 'onlineSavingsMsg'
Learn More

3-Methylpyrazole-4-carboxaldehyde 98.0+%, TCI America™

Catalog No. p-7133344
Change view
Click to view available options
Quantity:
1 g
200 mg
Catalog No. Quantity
M2643200MG 200 mg
M26431G 1 g
2 options

Catalog No. M2643200MG

Supplier: TCI America M2643200MG

Only null left

Chemical Identifiers

CAS 112758-40-4
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
MDL Number MFCD01313822
InChI Key NWDMGTFNIOCVDU-UHFFFAOYSA-N
Synonym 3-methyl-1h-pyrazole-4-carbaldehyde, 3-methyl-1h-pyrazole-4-carboxaldehyde, 3-methylpyrazole-4-carboxaldehyde, 3-methylpyrazole-4-carbaldehyde, 1h-pyrazole-4-carboxaldehyde, 3-methyl, 3-methyl-2h-pyrazole-4-carbaldehyde, 5-methylpyrazole-4-carbaldehyde, pubchem16065, acmc-1brby, 4-formyl-3-methylpyrazole
PubChem CID 2760056
IUPAC Name 5-methyl-1H-pyrazole-4-carbaldehyde
SMILES CC1=C(C=O)C=NN1

Specifications

Melting Point 110°C
Color White-Yellow
Quantity 200 mg
Formula Weight 110.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methylpyrazole-4-carboxaldehyde
TSCA No
Recommended Storage Refrigerator