missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methylpyrazole-4-carboxaldehyde 98.0+%, TCI America™
Supplier: TCI America M26431G
Specifications
| 3-Methylpyrazole-4-carboxaldehyde | |
| 110°C | |
| C5H6N2O | |
| 1 g | |
| NWDMGTFNIOCVDU-UHFFFAOYSA-N | |
| 5-methyl-1H-pyrazole-4-carbaldehyde | |
| 2760056 | |
| ≥98.0% (GC) |
| 112758-40-4 | |
| White-Yellow | |
| MFCD01313822 | |
| 3-methyl-1h-pyrazole-4-carbaldehyde, 3-methyl-1h-pyrazole-4-carboxaldehyde, 3-methylpyrazole-4-carboxaldehyde, 3-methylpyrazole-4-carbaldehyde, 1h-pyrazole-4-carboxaldehyde, 3-methyl, 3-methyl-2h-pyrazole-4-carbaldehyde, 5-methylpyrazole-4-carbaldehyde, pubchem16065, acmc-1brby, 4-formyl-3-methylpyrazole | |
| CC1=C(C=O)C=NN1 | |
| 110.12 | |
| 110.12 | |
| Crystalline Powder |
Chemical Identifiers
| 112758-40-4 | |
| 110.12 | |
| NWDMGTFNIOCVDU-UHFFFAOYSA-N | |
| 2760056 | |
| CC1=C(C=O)C=NN1 |
| C5H6N2O | |
| MFCD01313822 | |
| 3-methyl-1h-pyrazole-4-carbaldehyde, 3-methyl-1h-pyrazole-4-carboxaldehyde, 3-methylpyrazole-4-carboxaldehyde, 3-methylpyrazole-4-carbaldehyde, 1h-pyrazole-4-carboxaldehyde, 3-methyl, 3-methyl-2h-pyrazole-4-carbaldehyde, 5-methylpyrazole-4-carbaldehyde, pubchem16065, acmc-1brby, 4-formyl-3-methylpyrazole | |
| 5-methyl-1H-pyrazole-4-carbaldehyde |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator