missing translation for 'onlineSavingsMsg'
Learn More

3-Methylpiperidine 97.0+%, TCI America™

Catalog No. p-7134609
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
P044425ML 25 mL
P0444500ML 500 mL
2 options

Catalog No. P044425ML

Supplier: TCI America P044425ML

Only null left

Chemical Identifiers

CAS 626-56-2
Molecular Formula C6H13N
Molecular Weight (g/mol) 99.177
MDL Number MFCD00005994
InChI Key JEGMWWXJUXDNJN-UHFFFAOYSA-N
Synonym 3-pipecoline, piperidine, 3-methyl, beta-pipecoline, .beta.-pipecoline, 3-methyl-piperidine, .beta.-methylpiperidine, beta-methylpiperidine, 3-methyl piperidine, 3-pipecoline 8ci, 3ppc
PubChem CID 79081
IUPAC Name 3-methylpiperidine
SMILES CC1CCCNC1

Specifications

Color Yellow
Boiling Point 126°C
Quantity 25 mL
UN Number 1993
Formula Weight 99.18
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Methylpiperidine
EINECSNumber (5)-0768
TSCA Yes