missing translation for 'onlineSavingsMsg'
Learn More

3-Methylpiperidine 97.0+%, TCI America™

Catalog No. P044425ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P044425ML 25 mL
P0444500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P044425ML Supplier TCI America Supplier No. P044425ML

Chemical Identifiers

CAS 626-56-2
Molecular Formula C6H13N
Molecular Weight (g/mol) 99.177
MDL Number MFCD00005994
InChI Key JEGMWWXJUXDNJN-UHFFFAOYSA-N
Synonym 3-pipecoline, piperidine, 3-methyl, beta-pipecoline, .beta.-pipecoline, 3-methyl-piperidine, .beta.-methylpiperidine, beta-methylpiperidine, 3-methyl piperidine, 3-pipecoline 8ci, 3ppc
PubChem CID 79081
IUPAC Name 3-methylpiperidine
SMILES CC1CCCNC1

Specifications

Color Yellow
Boiling Point 126°C
Quantity 25 mL
UN Number 1993
Formula Weight 99.18
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Methylpiperidine
EINECSNumber (5)-0768
TSCA Yes