missing translation for 'onlineSavingsMsg'
Learn More

3-Methylphthalic Anhydride 96.0+%, TCI America™

Catalog No. M14641G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14641G 1 g
M14645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M14641G Supplier TCI America Supplier No. M14641G

Chemical Identifiers

CAS 4792-30-7
Molecular Formula C9H6O3
Molecular Weight (g/mol) 162.14
MDL Number MFCD00047316
InChI Key TWWAWPHAOPTQEU-UHFFFAOYSA-N
Synonym 3-methylphthalic anhydride, 4-methylisobenzofuran-1,3-dione, 1,3-isobenzofurandione, 4-methyl, 3-methyl phthalic anhydride, methylphthalic anhydride, phthalic anhydride, 3-methyl, 2,3-toluenedicarboxylic anhydride, 1,3-isobenzofurandione, methyl, 4-methyl-1,3-dihydro-2-benzofuran-1,3-dione, acmc-1arrb
PubChem CID 98500
IUPAC Name 4-methyl-1,3-dihydro-2-benzofuran-1,3-dione
SMILES CC1=CC=CC2=C1C(=O)OC2=O

Specifications

Melting Point 118°C
Quantity 1 g
Formula Weight 162.14
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methylphthalic Anhydride