accessibility menu, dialog, popup

UserName

CAS RN 4792-30-7

IUPAC Name: 4-methyl-1,3-dihydro-2-benzofuran-1,3-dione
Synonyms: 4-methyl-2-benzofuran-1,3-dione
Molecular Weight (g/mol): 162.14
Molecular Formula: C9H6O3
InChi Key: TWWAWPHAOPTQEU-UHFFFAOYSA-N
SMILES: CC1=CC=CC2=C1C(=O)OC2=O
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)