Learn More
3-Methylphthalic anhydride, 96%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 414780250
| Quantity | 25g |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C9H6O3 | |
| MFCD00047316 | |
| 3-methylphthalic anhydride, 4-methylisobenzofuran-1,3-dione, 1,3-isobenzofurandione, 4-methyl, 3-methyl phthalic anhydride, methylphthalic anhydride, phthalic anhydride, 3-methyl, 2,3-toluenedicarboxylic anhydride, 1,3-isobenzofurandione, methyl, 4-methyl-1,3-dihydro-2-benzofuran-1,3-dione, acmc-1arrb | |
| 4-methyl-2-benzofuran-1,3-dione |
Specifications
| 3-Methylphthalic anhydride | |
| 4792-30-7 | |
| 100.0 | |
| 96% | |
| C9H6O3 | |
| MFCD00047316 | |
| TWWAWPHAOPTQEU-UHFFFAOYSA-N | |
| 4-methyl-2-benzofuran-1,3-dione | |
| 98500 | |
| 96% |
| 96% | |
| 95.0 | |
| Authentic | |
| Glass bottle | |
| 25g | |
| 3-methylphthalic anhydride, 4-methylisobenzofuran-1,3-dione, 1,3-isobenzofurandione, 4-methyl, 3-methyl phthalic anhydride, methylphthalic anhydride, phthalic anhydride, 3-methyl, 2,3-toluenedicarboxylic anhydride, 1,3-isobenzofurandione, methyl, 4-methyl-1,3-dihydro-2-benzofuran-1,3-dione, acmc-1arrb | |
| CC1=CC=CC2=C1C(=O)OC2=O | |
| 162.14 | |
| 162.14 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respir
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minut
GHS Signal Word: Danger
EINECSNumber : 225-344-0
TSCA : TSCA
Promotions