missing translation for 'onlineSavingsMsg'
Learn More

3-Methylphenol, MilliporeSigma™ Supelco™

Catalog No. 111020622
Change view
Click to view available options
Quantity:
1000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020622 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020622 Supplier Merck Emd Millipore Supplier No. 442391

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

3-Methylphenol, a substituted phenol, is a common water pollutant. It is found to be toxic to fish and aquatic biota, even when present in trace quantities.

Agriculture and Environmental

Specifications

Packaging Ampule
Ignition Point 1036°F
CAS 108-39-4
Vapor Pressure <1 mmHg (20°C)
Vapor Density 3.72 (vs. air)
Density 1.034 g/mL (at 25°C)
Flash Point 86°C
Physical Form Neat
Linear Formula CH3C6H4 OH
MDL Number MFCD00002302
Melting Point 8°C to 10°C
Boiling Point 203°C
Synonym m-Cresol; 1-Hydroxy-3-methylbenzene; 3-Hydroxytoluene; 3-Methylphenol
Type Organic Pollutant Standard
Quantity 1000 mg
Formula Weight 108.14
Grade Analytical Standard
Show More Show Less