missing translation for 'onlineSavingsMsg'
Learn More

3-Methylpentane 99.0+%, TCI America™

Catalog No. M038325ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M038325ML 25 mL
M0383500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M038325ML Supplier TCI America Supplier No. M038325ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 96-14-0
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD00009342
InChI Key PFEOZHBOMNWTJB-UHFFFAOYSA-N
Synonym pentane, 3-methyl, unii-xd8o3ml76t, 3-methyl-pentane, xd8o3ml76t, 3-methylpentane, analytical standard, diethylmethylmethane, 3-methylpentyl, 3-methyl pentane, 3-methylpentan-3-yl, acmc-209s6l
PubChem CID 7282
IUPAC Name 3-methylpentane
SMILES CCC(C)CC

Specifications

Melting Point -118°C
Color Colorless
Quantity 25 mL
Physical Form Clear Liquid at 20°C
UN Number 1208
Boiling Point 63°C
Formula Weight 86.18
Percent Purity ≥99.0% (GC)
Chemical Name or Material 3-Methylpentane
EINECSNumber (2)-0006
RTECSNumber SA2995500
TSCA Yes