Learn More
3-Methylpentane, 99+%
CAS: 96-14-0 | C6H14 | 86.18 g/mol
Supplier: Thermo Scientific Chemicals L03133AP
Description
3-Methylpentane is used as a solvent in the preparation of vegetable oils, glues, coatings, and paints. It is also used in gasoline, rubber solvent, and petroleum ether.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Methylpentane is used as a solvent in the preparation of vegetable oils, glues, coatings, and paints. It is also used in gasoline, rubber solvent, and petroleum ether.
Solubility
Soluble in water. 0.013 g/L at 20°C). Miscible in ether, acetone. Soluble in ethanol.
Notes
Store in a cool, dark and well-ventilated place. Store away from incompatible materials such as oxidizing agents.
Specifications
| 3-Methylpentane | |
| -118°C | |
| 63°C to 65°C | |
| Petroleum-like | |
| 1.377 | |
| 500 mL | |
| 1730734 | |
| Soluble in water. 0.013g/L at 20°C). Miscible in ether,acetone. Soluble in ethanol. | |
| CCC(C)CC | |
| 86.18 | |
| 86.18 |
| 96-14-0 | |
| 0.663 | |
| −32°C (−26°F) | |
| C6H14 | |
| MFCD00009342 | |
| UN1208 | |
| pentane, 3-methyl, unii-xd8o3ml76t, 3-methyl-pentane, xd8o3ml76t, 3-methylpentane, analytical standard, diethylmethylmethane, 3-methylpentyl, 3-methyl pentane, 3-methylpentan-3-yl, acmc-209s6l | |
| PFEOZHBOMNWTJB-UHFFFAOYSA-N | |
| 3-methylpentane | |
| 7282 | |
| ≥99% |
Chemical Identifiers
| 96-14-0 | |
| 86.18 | |
| PFEOZHBOMNWTJB-UHFFFAOYSA-N | |
| 7282 | |
| CCC(C)CC |
| C6H14 | |
| MFCD00009342 | |
| pentane, 3-methyl, unii-xd8o3ml76t, 3-methyl-pentane, xd8o3ml76t, 3-methylpentane, analytical standard, diethylmethylmethane, 3-methylpentyl, 3-methyl pentane, 3-methylpentan-3-yl, acmc-209s6l | |
| 3-methylpentane |
Safety and Handling
GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
H225-H304-H315-H319-H336
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEXANES
EINECSNumber : 202-481-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only