missing translation for 'onlineSavingsMsg'
Learn More

3-Methylpentane, 99+%

CAS: 96-14-0 | C6H14 | 86.18 g/mol

Supplier:  Thermo Scientific Chemicals L03133AE

 View more versions of this product

Catalog No. AAL03133AE


Only null left
Explore available promotions

Description

Description

3-Methylpentane is used as a solvent in the preparation of vegetable oils, glues, coatings, and paints. It is also used in gasoline, rubber solvent, and petroleum ether.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methylpentane is used as a solvent in the preparation of vegetable oils, glues, coatings, and paints. It is also used in gasoline, rubber solvent, and petroleum ether.

Solubility
Soluble in water. 0.013 g/L at 20°C). Miscible in ether, acetone. Soluble in ethanol.

Notes
Store in a cool, dark and well-ventilated place. Store away from incompatible materials such as oxidizing agents.
Specifications

Specifications

3-Methylpentane
-118°C
63°C to 65°C
Petroleum-like
1.377
100 mL
1730734
Soluble in water. 0.013g/L at 20°C). Miscible in ether,acetone. Soluble in ethanol.
CCC(C)CC
86.18
86.18
96-14-0
0.663
−32°C (−26°F)
C6H14
MFCD00009342
UN1208
pentane, 3-methyl, unii-xd8o3ml76t, 3-methyl-pentane, xd8o3ml76t, 3-methylpentane, analytical standard, diethylmethylmethane, 3-methylpentyl, 3-methyl pentane, 3-methylpentan-3-yl, acmc-209s6l
PFEOZHBOMNWTJB-UHFFFAOYSA-N
3-methylpentane
7282
≥99%

Chemical Identifiers

96-14-0
86.18
PFEOZHBOMNWTJB-UHFFFAOYSA-N
7282
CCC(C)CC
C6H14
MFCD00009342
pentane, 3-methyl, unii-xd8o3ml76t, 3-methyl-pentane, xd8o3ml76t, 3-methylpentane, analytical standard, diethylmethylmethane, 3-methylpentyl, 3-methyl pentane, 3-methylpentan-3-yl, acmc-209s6l
3-methylpentane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c

H225-H304-H315-H319-H336

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEXANES

EINECSNumber : 202-481-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only