missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(3-Methylisoxazol-5-ylmethyl)amine, 97%, Thermo Scientific™
$251.06 - $251.06
Chemical Identifiers
| CAS | 154016-55-4 |
|---|---|
| Molecular Formula | C5H8N2O |
| Molecular Weight (g/mol) | 112.132 |
| MDL Number | MFCD06738858 |
| InChI Key | JKVMPILAJBLISV-UHFFFAOYSA-N |
| Synonym | 3-methylisoxazol-5-ylmethyl amine, 3-methylisoxazol-5-yl methanamine, 5-aminomethyl-3-methylisoxazole, 3-methyl-1,2-oxazol-5-yl methanamine, 3-methylisoxazol-5-yl methylamine, 1-3-methylisoxazol-5-yl methanamine, 5-isoxazolemethanamine, 3-methyl, 1-3-methyl-1,2-oxazol-5-yl methanamine, 1-3-methyl-5-isoxazolyl methanamine, c-3-methyl-isoxazol-5-yl-methylamine |
| PubChem CID | 16481072 |
| IUPAC Name | (3-methyl-1,2-oxazol-5-yl)methanamine |
| SMILES | CC1=NOC(=C1)CN |
Chemical Identifiers
| 154016-55-4 | |
| 112.132 | |
| JKVMPILAJBLISV-UHFFFAOYSA-N | |
| 16481072 | |
| CC1=NOC(=C1)CN |
| C5H8N2O | |
| MFCD06738858 | |
| 3-methylisoxazol-5-ylmethyl amine, 3-methylisoxazol-5-yl methanamine, 5-aminomethyl-3-methylisoxazole, 3-methyl-1,2-oxazol-5-yl methanamine, 3-methylisoxazol-5-yl methylamine, 1-3-methylisoxazol-5-yl methanamine, 5-isoxazolemethanamine, 3-methyl, 1-3-methyl-1,2-oxazol-5-yl methanamine, 1-3-methyl-5-isoxazolyl methanamine, c-3-methyl-isoxazol-5-yl-methylamine | |
| (3-methyl-1,2-oxazol-5-yl)methanamine |
Specifications
| 154016-55-4 | |
| 100.0 | |
| Amber Glass Bottle | |
| MFCD06738858 | |
| 3-methylisoxazol-5-ylmethyl amine, 3-methylisoxazol-5-yl methanamine, 5-aminomethyl-3-methylisoxazole, 3-methyl-1,2-oxazol-5-yl methanamine, 3-methylisoxazol-5-yl methylamine, 1-3-methylisoxazol-5-yl methanamine, 5-isoxazolemethanamine, 3-methyl, 1-3-methyl-1,2-oxazol-5-yl methanamine, 1-3-methyl-5-isoxazolyl methanamine, c-3-methyl-isoxazol-5-yl-methylamine | |
| CC1=NOC(=C1)CN | |
| 112.132 | |
| 112.13 | |
| (3-Methylisoxazol-5-ylmethyl)amine |
| 96.5 | |
| 97% | |
| C5H8N2O | |
| 250 mg | |
| JKVMPILAJBLISV-UHFFFAOYSA-N | |
| (3-methyl-1,2-oxazol-5-yl)methanamine | |
| 16481072 | |
| 97% |
Safety and Handling
GHS P Statement Harmful if inhaled. Causes severe skin burns and eye damage. Harmful if swallowed. Causes serious eye damage. Harmful in contact with skin.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Immediately call a POISON CENTER or doctor/physician. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cau
Danger