CAS RN 154016-55-4
IUPAC Name:
1-(3-methyl-1,2-oxazol-5-yl)methanamine
Synonyms:
1-(3-methyl-1,2-oxazol-5-yl)methanamine
Molecular Weight (g/mol):
112.13
Molecular Formula:
C5H8N2O
InChi Key:
JKVMPILAJBLISV-UHFFFAOYSA-N
SMILES:
CC1=NOC(CN)=C1
1
–
2
of
2
results
2
(3-Methylisoxazol-5-ylmethyl)amine, 97%, Thermo Scientific™
CAS: 154016-55-4 Molecular Formula: C5H8N2O Molecular Weight (g/mol): 112.132 MDL Number: MFCD06738858 InChI Key: JKVMPILAJBLISV-UHFFFAOYSA-N Synonym: 3-methylisoxazol-5-ylmethyl amine,3-methylisoxazol-5-yl methanamine,5-aminomethyl-3-methylisoxazole,3-methyl-1,2-oxazol-5-yl methanamine,3-methylisoxazol-5-yl methylamine,1-3-methylisoxazol-5-yl methanamine,5-isoxazolemethanamine, 3-methyl,1-3-methyl-1,2-oxazol-5-yl methanamine,1-3-methyl-5-isoxazolyl methanamine,c-3-methyl-isoxazol-5-yl-methylamine PubChem CID: 16481072 IUPAC Name: (3-methyl-1,2-oxazol-5-yl)methanamine SMILES: CC1=NOC(=C1)CN