missing translation for 'onlineSavingsMsg'
Learn More

(3-Methylisoxazol-5-ylmethyl)amine, 97%, Thermo Scientific™

Catalog No. cc27013cb
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC27013CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC27013CB Supplier Thermo Scientific Chemicals Supplier No. CC27013CB
Only null left

Chemical Identifiers

CAS 154016-55-4
Molecular Formula C5H8N2O
Molecular Weight (g/mol) 112.132
MDL Number MFCD06738858
InChI Key JKVMPILAJBLISV-UHFFFAOYSA-N
Synonym 3-methylisoxazol-5-ylmethyl amine, 3-methylisoxazol-5-yl methanamine, 5-aminomethyl-3-methylisoxazole, 3-methyl-1,2-oxazol-5-yl methanamine, 3-methylisoxazol-5-yl methylamine, 1-3-methylisoxazol-5-yl methanamine, 5-isoxazolemethanamine, 3-methyl, 1-3-methyl-1,2-oxazol-5-yl methanamine, 1-3-methyl-5-isoxazolyl methanamine, c-3-methyl-isoxazol-5-yl-methylamine
PubChem CID 16481072
IUPAC Name (3-methyl-1,2-oxazol-5-yl)methanamine
SMILES CC1=NOC(=C1)CN

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 112.13
Percent Purity 97%
Chemical Name or Material (3-Methylisoxazol-5-ylmethyl)amine
Hazard Category Danger
Hazard Statement GHS P Statement Harmful if inhaled. Causes severe skin burns and eye damage. Harmful if swallowed. Causes serious eye damage. Harmful in contact with skin.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Immediately call a POISON CENTER or doctor/physician. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cau