missing translation for 'onlineSavingsMsg'
Learn More

3-Methylindole 98.0+%, TCI America™

Catalog No. M03475G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M03475G 5 g
M03471G 1 g
M034725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M03475G Supplier TCI America Supplier No. M03475G

Chemical Identifiers

CAS 83-34-1
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.178
MDL Number MFCD00005627
InChI Key ZFRKQXVRDFCRJG-UHFFFAOYSA-N
Synonym 3-methylindole, skatole, scatole, skatol, 1h-indole, 3-methyl, beta-methylindole, indole, 3-methyl, 3-mi, 3-methyl-4,5-benzopyrrole, 3-methyl indole
PubChem CID 6736
ChEBI CHEBI:9171
IUPAC Name 3-methyl-1H-indole
SMILES CC1=CNC2=CC=CC=C12

Specifications

Melting Point 96°C
Color White-Yellow
Boiling Point 141°C
Quantity 5 g
Formula Weight 131.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methylindole
RTECSNumber NM0350000
TSCA Yes