accessibility menu, dialog, popup

UserName

H3CHN

CAS RN 83-34-1

IUPAC Name: 3-methyl-1H-indole
Synonyms: scatole 3-methyl-4,5-benzopyrrole 3MI skatole (substance) methyl-1h-indole
Molecular Weight (g/mol): 131.18
Molecular Formula: C9H9N
InChi Key: ZFRKQXVRDFCRJG-UHFFFAOYSA-N
SMILES: CC1=CNC2=CC=CC=C12
Boiling Point 
  • (3)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (5)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)