missing translation for 'onlineSavingsMsg'
Learn More

3-Methylindole, 99%

Catalog No. p-7043987
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0389006 5 g
AAL0389014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0389006 Supplier Thermo Scientific Chemicals Supplier No. L0389006
Only null left

CAS: 83-34-1 | C9H9N | 131.178 g/mol

3-Methylindole is often used as a component of commercial fragrances and perfumes.It helps fluorescent guanosine analogue, which in a dimethoxytrityl, phosphoramidite protected form, can be site-specifically inserted into oligonucleotides through a 3?5?phosphodiester linkage using an automated DNA synthesizer

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methylindole is often used as a component of commercial fragrances and perfumes.It helps fluorescent guanosine analogue, which in a dimethoxytrityl, phosphoramidite protected form, can be site-specifically inserted into oligonucleotides through a 3?5?phosphodiester linkage using an automated DNA synthesizer

Solubility
Soluble in water, Ether, Alcohols, Benzene, Acetone, Chloroform.

Notes
Light Sensitive. Store in dark away from oxidizing agents. Incompatible with light, strong acids, acid ahydrides, acid chlorides.

Chemical Identifiers

CAS 83-34-1
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.178
MDL Number MFCD00005627
InChI Key ZFRKQXVRDFCRJG-UHFFFAOYSA-N
Synonym 3-methylindole, skatole, scatole, skatol, 1h-indole, 3-methyl, beta-methylindole, indole, 3-methyl, 3-mi, 3-methyl-4,5-benzopyrrole, 3-methyl indole
PubChem CID 6736
ChEBI CHEBI:9171
IUPAC Name 3-methyl-1H-indole
SMILES CC1=CNC2=CC=CC=C12

Specifications

Melting Point 94°C to 97°C
Boiling Point 265°C to 268°C
Flash Point 132°C (269°F)
Odor Stench
Quantity 5 g
Beilstein 111296
Sensitivity Light sensitive
Merck Index 14,8560
Solubility Information Soluble in water,Ether,Alcohols,Benzene,Acetone,Chloroform.
Formula Weight 131.18
Percent Purity 99%
Chemical Name or Material 3-Methylindole
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 201-471-7
RTECSNumber NM0350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only