missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methylhistamine dihydrochloride, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Packaging | Poly Bottle |
| CAS | 36475-47-5 |
| Physical Form | Neat |
| Linear Formula | C6H11N3 · 2 HCl |
| MDL Number | MFCD00037002 |
| Synonym | 3-Methyl-4-(beta-aminoethyl)imidazole |
| Type | Analytical Standard |
| Quantity | 50 mg |
| Formula Weight | 198.09 |
| Grade | Analytical Standard |