missing translation for 'onlineSavingsMsg'
Learn More

3-Methylhippuric acid, 97%

Catalog No. AAH5345806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5345806 5 g
AAH5345803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5345806 Supplier Thermo Scientific Chemicals Supplier No. H5345806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 27115-49-7 | C10H11NO3 | 193.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27115-49-7
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.20
MDL Number MFCD00044399
InChI Key YKAKNMHEIJUKEX-UHFFFAOYSA-N
Synonym 3-methylhippuric acid, m-methylhippuric acid, n-m-toluoyl glycine, m-toluric acid, n-3-methylbenzoyl glycine, 3-methylbenzoylglycine, glycine, n-3-methylbenzoyl, meta-methylhippuric acid, hippuric acid, m-methyl, 3-methyl hippuric acid
PubChem CID 99223
ChEBI CHEBI:68500
SMILES CC1=CC=CC(=C1)C(=O)NCC(O)=O

Specifications

Melting Point 138°C to 140°C
Quantity 5 g
IUPAC Name 2-[(3-methylbenzoyl)amino]acetic acid
Formula Weight 193.2
Percent Purity 97%
Chemical Name or Material 3-Methylhippuric acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only