missing translation for 'onlineSavingsMsg'
Learn More

3-Methylhexyl Acetate 95.0+%, TCI America™

Catalog No. A13961G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13961G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A13961G Supplier TCI America Supplier No. A13961G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 50373-54-1
Molecular Formula C9H18O2
Molecular Weight (g/mol) 158.241
MDL Number MFCD01321123
InChI Key JASARFBWVJNEMI-UHFFFAOYSA-N
Synonym Acetic Acid 3-Methylhexyl Ester
PubChem CID 44630061
IUPAC Name 3-methylhexyl acetate
SMILES CCCC(C)CCOC(=O)C

Specifications

Color Colorless
Boiling Point 184°C
Quantity 1 g
UN Number 3272
Formula Weight 158.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methylhexyl Acetate
EINECSNumber (2)-0735
TSCA No