accessibility menu, dialog, popup

UserName

CAS RN 50373-54-1

IUPAC Name: 3-methylhexyl acetate
Synonyms: 3-methylhexyl acetate
Molecular Weight (g/mol): 158.24
Molecular Formula: C9H18O2
InChi Key: JASARFBWVJNEMI-UHFFFAOYNA-N
SMILES: CCCC(C)CCOC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results