missing translation for 'onlineSavingsMsg'
Learn More

3-Methylhexane 95.0+%, TCI America™

Catalog No. p-7132108
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M03405ML 5 mL
M034025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M03405ML Supplier TCI America Supplier No. M03405ML
Only null left

Chemical Identifiers

CAS 589-34-4
Molecular Formula C7H16
Molecular Weight (g/mol) 100.205
MDL Number MFCD00009408
InChI Key VLJXXKKOSFGPHI-UHFFFAOYSA-N
Synonym hexane, 3-methyl, 2-ethylpentane, 3-methyl hexane, 3-methyl-hexane, 3-methylhexane, dsstox_cid_8974, acmc-209m8a, s-+-3-methylhexane, dsstox_rid_80156, dsstox_gsid_44334
PubChem CID 11507
IUPAC Name 3-methylhexane
SMILES CCCC(C)CC

Specifications

Color Colorless
Boiling Point 91°C
Quantity 5 mL
UN Number 1206
Formula Weight 100.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methylhexane
EINECSNumber (2)-0007
TSCA Yes