accessibility menu, dialog, popup

UserName

CAS RN 589-34-4

H3CCH3CH3

CAS RN 589-34-4

IUPAC Name: 3-methylhexane
Synonyms: 3-methylhexane
Molecular Weight (g/mol): 100.21
Molecular Formula: C7H16
InChi Key: VLJXXKKOSFGPHI-UHFFFAOYNA-N
SMILES: CCCC(C)CC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results