missing translation for 'onlineSavingsMsg'
Learn More

3-Methylheptane 97.0+%, TCI America™

Catalog No. M03341ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M03341ML 1 mL
M03345ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M03341ML Supplier TCI America Supplier No. M03341ML

Chemical Identifiers

CAS 589-81-1
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00027244
InChI Key LAIUFBWHERIJIH-UHFFFAOYSA-N
Synonym heptane, 3-methyl, 2-ethylhexane, 3-methyl-heptane, 2-butylbutane, 3-methyl-heptan, heptane, 3-methyl-, r, acmc-20mdw0, 3-methyl-s-heptane, 3-methylheptane, acmc-1axq2
PubChem CID 11519
IUPAC Name 3-methylheptane
SMILES CCCCC(C)CC

Specifications

Melting Point -121°C
Color Colorless
Boiling Point 119°C
Quantity 1 mL
UN Number 1262
Formula Weight 114.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methylheptane
EINECSNumber (2)-0008
TSCA Yes