missing translation for 'onlineSavingsMsg'
Learn More

3-Methylheptane, 97%

Catalog No. p-7045793
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1200603 1 g
AAL1200606 5 g
AAL1200614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1200603 Supplier Thermo Scientific Chemicals Supplier No. L1200603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 589-81-1 | C8H18 | 114.232 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 589-81-1
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00027244
InChI Key LAIUFBWHERIJIH-UHFFFAOYSA-N
Synonym heptane, 3-methyl, 2-ethylhexane, 3-methyl-heptane, 2-butylbutane, 3-methyl-heptan, heptane, 3-methyl-, r, acmc-20mdw0, 3-methyl-s-heptane, 3-methylheptane, acmc-1axq2
PubChem CID 11519
IUPAC Name 3-methylheptane
SMILES CCCCC(C)CC

Specifications

Melting Point -121°C
Density 0.71
Boiling Point 118°C to 120°C
Flash Point 7°C (44°F)
Refractive Index 1.398
Quantity 1 g
UN Number UN1262
Beilstein 1730777
Formula Weight 114.23
Percent Purity 97%
Chemical Name or Material 3-Methylheptane
Show More Show Less
Hazard Category H225-H304-H315-H319-H336
Hazard Statement GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: OCTANES
EINECSNumber 209-660-6
TSCA Yes

RUO – Research Use Only