missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methylglutaric Acid 99.0+%, TCI America™
$154.71 - $415.68
Chemical Identifiers
| CAS | 626-51-7 |
|---|---|
| Molecular Formula | C6H10O4 |
| Molecular Weight (g/mol) | 146.142 |
| MDL Number | MFCD00002727 |
| InChI Key | XJMMNTGIMDZPMU-UHFFFAOYSA-N |
| Synonym | 3-methylglutaric acid, methylglutaric acid, pentanedioic acid, 3-methyl, 3-methyl-pentanedioic acid, 3-methylglutaricacid, beta-methylglutaric acid, unii-3q0p190c7b, 3-methylpentanedioate, b-methylglutarate, 3-methylglutarate |
| PubChem CID | 12284 |
| ChEBI | CHEBI:68566 |
| IUPAC Name | 3-methylpentanedioic acid |
| SMILES | CC(CC(=O)O)CC(=O)O |
Chemical Identifiers
| 626-51-7 | |
| 146.142 | |
| XJMMNTGIMDZPMU-UHFFFAOYSA-N | |
| 12284 | |
| 3-methylpentanedioic acid |
| C6H10O4 | |
| MFCD00002727 | |
| 3-methylglutaric acid, methylglutaric acid, pentanedioic acid, 3-methyl, 3-methyl-pentanedioic acid, 3-methylglutaricacid, beta-methylglutaric acid, unii-3q0p190c7b, 3-methylpentanedioate, b-methylglutarate, 3-methylglutarate | |
| CHEBI:68566 | |
| CC(CC(=O)O)CC(=O)O |
Specifications
| 626-51-7 | |
| White | |
| MFCD00002727 | |
| 3-methylglutaric acid, methylglutaric acid, pentanedioic acid, 3-methyl, 3-methyl-pentanedioic acid, 3-methylglutaricacid, beta-methylglutaric acid, unii-3q0p190c7b, 3-methylpentanedioate, b-methylglutarate, 3-methylglutarate | |
| CC(CC(=O)O)CC(=O)O | |
| 146.142 | |
| CHEBI:68566 | |
| ≥99.0% (T) | |
| 3-Methylglutaric Acid |
| 87°C | |
| C6H10O4 | |
| 5 g | |
| XJMMNTGIMDZPMU-UHFFFAOYSA-N | |
| 3-methylpentanedioic acid | |
| 12284 | |
| 146.14 | |
| Crystalline Powder |
Safety and Handling
TSCA : No