missing translation for 'onlineSavingsMsg'
Learn More

3-Methylglutaric Acid 99.0+%, TCI America™

Supplier:  TCI America M033025G

 View more versions of this product

Catalog No. M033025G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Methylglutaric Acid
87°C
C6H10O4
25 g
XJMMNTGIMDZPMU-UHFFFAOYSA-N
3-methylpentanedioic acid
12284
146.14
Crystalline Powder
626-51-7
White
MFCD00002727
3-methylglutaric acid, methylglutaric acid, pentanedioic acid, 3-methyl, 3-methyl-pentanedioic acid, 3-methylglutaricacid, beta-methylglutaric acid, unii-3q0p190c7b, 3-methylpentanedioate, b-methylglutarate, 3-methylglutarate
CC(CC(=O)O)CC(=O)O
146.142
CHEBI:68566
≥99.0% (T)

Chemical Identifiers

626-51-7
146.142
XJMMNTGIMDZPMU-UHFFFAOYSA-N
12284
3-methylpentanedioic acid
C6H10O4
MFCD00002727
3-methylglutaric acid, methylglutaric acid, pentanedioic acid, 3-methyl, 3-methyl-pentanedioic acid, 3-methylglutaricacid, beta-methylglutaric acid, unii-3q0p190c7b, 3-methylpentanedioate, b-methylglutarate, 3-methylglutarate
CHEBI:68566
CC(CC(=O)O)CC(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions