missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methylglutaconic acid, Mixture of E and Z Isomers, ≥98.0% (HPLC), MilliporeSigma™ Supelco™
$1131.97 - $1131.97
Chemical Identifiers
| MDL Number | MFCD01556044 |
|---|---|
| Synonym | 3-Methyl-2-pentenedioic acid; NSC 249232 |
Chemical Identifiers
| MFCD01556044 | |
| 3-Methyl-2-pentenedioic acid; NSC 249232 |
Specifications
| 5746-90-7 | |
| MFCD01556044 | |
| 3-Methyl-2-pentenedioic acid; NSC 249232 | |
| ≥98.0% (HPLC) |
| C6H8O4 | |
| 250 mg | |
| 144.13 |